Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 15 - 16 | 2282 | 109 | |
XM_012100634.2 | XP_011956024.1 | 11 - 12 | 1517 | 109 | ||
XM_027958477.1 | XP_027814278.1 | 14 - 15 | 2254 | 109 | ||
XM_027958478.1 | XP_027814279.1 | 16 - 17 | 2186 | 109 |

More Information
Set Membership: |
![]() ![]() |
