Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 19 - 20 | 3142 | 73 | |
XM_012100634.2 | XP_011956024.1 | 15 - 16 | 2377 | 73 | ||
XM_027958477.1 | XP_027814278.1 | 18 - 19 | 3114 | 73 | ||
XM_027958478.1 | XP_027814279.1 | 20 - 21 | 3046 | 73 |
