Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 23 - 24 | 3730 | 77 | |
XM_012100634.2 | XP_011956024.1 | 19 - 20 | 2965 | 77 | ||
XM_027958477.1 | XP_027814278.1 | 22 - 23 | 3702 | 77 | ||
XM_027958478.1 | XP_027814279.1 | 24 - 25 | 3634 | 77 |
