Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 24 - 25 | 3883 | 83 | |
XM_012100634.2 | XP_011956024.1 | 20 - 21 | 3118 | 83 | ||
XM_027958477.1 | XP_027814278.1 | 23 - 24 | 3855 | 83 | ||
XM_027958478.1 | XP_027814279.1 | 25 - 26 | 3787 | 83 |
