Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 25 - 26 | 4084 | 72 | |
XM_012100634.2 | XP_011956024.1 | 21 - 22 | 3319 | 72 | ||
XM_027958477.1 | XP_027814278.1 | 24 - 25 | 4056 | 72 | ||
XM_027958478.1 | XP_027814279.1 | 26 - 27 | 3988 | 72 |
