Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 3 - 4 | 312 | 81 | |
XM_027958477.1 | XP_027814278.1 | 2 - 3 | 284 | 81 | ||
XM_027958478.1 | XP_027814279.1 | 3 - 4 | 312 | 81 |
