Assay Details
| Gene Symbol: ZNF394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021046.4 | XP_004021095.2 | 1 - 2 | 793 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF394| Gene Symbol: ZNF394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021046.4 | XP_004021095.2 | 1 - 2 | 793 | 80 | |
Set Membership: |
|