Assay Details
| Gene Symbol: CTPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012108068.3 | XP_011963458.2 | 10 - 11 | 3202 | 61 | |
| XM_012108163.2 | XP_011963553.2 | 7 - 8 | 749 | 61 | ||
| XM_015091894.2 | XP_014947380.2 | 10 - 11 | 1225 | 61 | ||
| XM_015091895.2 | XP_014947381.1 | 10 - 11 | 1225 | 61 | ||
More Information
Set Membership: |
|

CTPS1
Genomic Map