Assay Details
| Gene Symbol: EFNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009108.3 | XP_004009157.1 | 2 - 3 | 730 | 59 | |
| XM_012179028.2 | XP_012034418.1 | 2 - 3 | 730 | 59 | ||
More Information
Set Membership: |
|

EFNA5
Genomic Map