Assay Details
Gene Symbol: CERS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969740.1 | XP_027825541.1 | 9 - 10 | 989 | 59 | |
XM_027969741.1 | XP_027825542.1 | 10 - 11 | 1099 | 59 | ||
XM_027969742.1 | XP_027825543.1 | 9 - 10 | 1055 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
