Assay Details
| Gene Symbol: RAB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016962.4 | XP_004017011.1 | 4 - 5 | 618 | 103 | |
| XM_012096890.3 | XP_011952280.1 | 4 - 5 | 815 | 103 | ||
More Information
Set Membership: |
|

RAB3C
Genomic Map