Assay Details
Gene Symbol: TRIQK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012184145.3 | XP_012039535.1 | 4 - 5 | 407 | 65 | |
XM_012184146.3 | XP_012039536.1 | 3 - 4 | 241 | 65 | ||
XM_027973234.1 | XP_027829035.1 | 4 - 5 | 404 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
