Assay Details
| Gene Symbol: PIGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106260.3 | XP_011961650.1 | 4 - 5 | 1118 | 65 | |
| XM_027963637.1 | XP_027819438.1 | 4 - 5 | 1118 | 65 | ||
More Information
Set Membership: |
|

PIGA
Genomic Map