Assay Details
Gene Symbol: LOC114113113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964796.1 | XP_027820597.1 | 3 - 4 | 301 | 103 |
Gene Symbol: LOC101122080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015093334.2 | XP_014948820.1 | 8 - 9 | 746 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
