Assay Details
| Gene Symbol: AAAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006301.4 | XP_004006350.2 | 14 - 15 | 1465 | 61 | |
| XM_012174192.3 | XP_012029582.2 | 15 - 16 | 1489 | 61 | ||
More Information
Set Membership: |
|

AAAS
Genomic Map