Assay Details
Gene Symbol: VCPIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011714.4 | XP_004011763.2 | 2 - 3 | 2798 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
