Assay Details
Gene Symbol: LOC101111607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004928.4 | XP_004004977.1 | 3 - 4 | 728 | 67 | |
XM_012146300.3 | XP_012001690.1 | 3 - 4 | 645 | 67 | ||
XM_012146310.3 | XP_012001700.1 | 3 - 4 | 795 | 67 | ||
XM_027965189.1 | XP_027820990.1 | 3 - 4 | 572 | 67 | ||
XM_027965190.1 | XP_027820991.1 | 3 - 4 | 675 | 67 | ||
XM_027965192.1 | XP_027820993.1 | 3 - 4 | 539 | 67 |
