Assay Details
Gene Symbol: LOC101111607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004928.4 | XP_004004977.1 | 4 - 5 | 935 | 72 | |
XM_012146300.3 | XP_012001690.1 | 4 - 5 | 852 | 72 | ||
XM_012146310.3 | XP_012001700.1 | 4 - 5 | 1002 | 72 | ||
XM_027965189.1 | XP_027820990.1 | 4 - 5 | 779 | 72 | ||
XM_027965190.1 | XP_027820991.1 | 4 - 5 | 882 | 72 | ||
XM_027965192.1 | XP_027820993.1 | 4 - 5 | 746 | 72 |
