Assay Details
| Gene Symbol: LOC101117682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012902.4 | XP_004012951.2 | 1 - 2 | 488 | 74 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117682| Gene Symbol: LOC101117682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012902.4 | XP_004012951.2 | 1 - 2 | 488 | 74 | |