Assay Details
Gene Symbol: LRRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957731.1 | XP_027813532.1 | 2 - 3 | 283 | 69 | |
XM_027957732.1 | XP_027813533.1 | 2 - 3 | 264 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
