Assay Details
Gene Symbol: ZNF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012105052.3 | XP_011960442.2 | 2 - 3 | 2069 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
