Assay Details
| Gene Symbol: ZNF25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012105052.3 | XP_011960442.2 | 4 - 5 | 2233 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF25| Gene Symbol: ZNF25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012105052.3 | XP_011960442.2 | 4 - 5 | 2233 | 113 | |
Set Membership: |
|