Assay Details
| Gene Symbol: IL13RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963636.1 | XP_027819437.1 | 3 - 4 | 418 | 72 | |
| GenBank mRNA | AY377582.1 | - | 2 - 3 | 287 | 72 | |
More Information
Set Membership: |
|

IL13RA1
Genomic Map