Assay Details
| Gene Symbol: TFE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022095.3 | XP_004022144.1 | 8 - 9 | 1375 | 57 | |
| XM_012106728.3 | XP_011962118.1 | 7 - 8 | 1451 | 57 | ||
| XM_027962861.1 | XP_027818662.1 | 6 - 7 | 3318 | 57 | ||
| XM_027962862.1 | XP_027818663.1 | 6 - 7 | 3272 | 57 | ||
More Information
Set Membership: |
|

TFE3
Genomic Map