Assay Details
| Gene Symbol: SUOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006596.4 | XP_004006645.2 | 3 - 4 | 511 | 64 | |
| XM_012174764.2 | XP_012030154.2 | 3 - 4 | 348 | 64 | ||
More Information
Set Membership: |
|

SUOX
Genomic Map