Assay Details
| Gene Symbol: TENT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097305.2 | XP_014952791.2 | 2 - 3 | 808 | 100 | |
| XM_027972307.1 | XP_027828108.1 | 2 - 3 | 752 | 100 | ||
More Information
Set Membership: |
|

TENT5A
Genomic Map