Assay Details
Gene Symbol: NR4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012120268.2 | XP_011975658.2 | 5 - 6 | 2525 | 68 | |
XM_027964393.1 | XP_027820194.1 | 7 - 8 | 2341 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
