Assay Details
Gene Symbol: MRPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018184.3 | XP_004018233.1 | 3 - 4 | 358 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
