Assay Details
Gene Symbol: LOC101120701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018866.4 | XP_004018915.1 | 7 - 8 | 1195 | 103 | |
XM_027958406.1 | XP_027814207.1 | 7 - 8 | 1196 | 103 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
