Assay Details
Gene Symbol: LOC101121385 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092152.2 | XP_014947638.2 | 8 - 9 | 936 | 82 | |
XM_027964822.1 | XP_027820623.1 | 8 - 9 | 930 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
