Assay Details
Gene Symbol: LRFN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977395.1 | XP_027833196.1 | 3 - 4 | 2579 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
