Assay Details
Gene Symbol: LOC114110575 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961443.1 | XP_027817244.1 | 3 - 4 | 346 | 77 |
Gene Symbol: LOC101114404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979709.1 | XP_027835510.1 | 16 - 17 | 2359 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
