Assay Details
| Gene Symbol: LOC101114404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979709.1 | XP_027835510.1 | 5 - 6 | 640 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101114404| Gene Symbol: LOC101114404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979709.1 | XP_027835510.1 | 5 - 6 | 640 | 116 | |
Set Membership: |
|