Assay Details
Gene Symbol: STX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014435.3 | XP_004014484.1 | 4 - 5 | 1114 | 65 | |
XM_004014436.3 | XP_004014485.1 | 4 - 5 | 1166 | 65 | ||
XM_004014437.3 | XP_004014486.1 | 3 - 4 | 1154 | 65 | ||
XM_004014438.3 | XP_004014487.1 | 3 - 4 | 1102 | 65 | ||
XM_012188916.2 | XP_012044306.1 | 5 - 6 | 519 | 65 | ||
XM_012188917.2 | XP_012044307.1 | 5 - 6 | 455 | 65 | ||
XM_012188918.2 | XP_012044308.1 | 4 - 5 | 507 | 65 | ||
XM_027977007.1 | XP_027832808.1 | 3 - 4 | 353 | 65 | ||
XM_027977008.1 | XP_027832809.1 | 5 - 6 | 519 | 65 | ||
XM_027977010.1 | XP_027832811.1 | 5 - 6 | 390 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
