Assay Details
| Gene Symbol: LOC101111633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180022.3 | XP_012035412.2 | 3 - 4 | 1646 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101111633| Gene Symbol: LOC101111633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180022.3 | XP_012035412.2 | 3 - 4 | 1646 | 104 | |
Set Membership: |
|