Assay Details
| Gene Symbol: LOC101117224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973853.1 | XP_027829654.1 | 33 - 34 | 5227 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117224| Gene Symbol: LOC101117224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973853.1 | XP_027829654.1 | 33 - 34 | 5227 | 108 | |
Set Membership: |
|