Assay Details
Gene Symbol: LOC114111341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963108.1 | XP_027818909.1 | 1 - 2 | 184 | 71 |
Gene Symbol: LOC101117224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973853.1 | XP_027829654.1 | 44 - 45 | 6586 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
