Assay Details
| Gene Symbol: SLC23A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004939.3 | XP_004004988.1 | 11 - 12 | 1575 | 55 | |
| XM_012146424.2 | XP_012001814.1 | 10 - 11 | 1191 | 55 | ||
More Information
Set Membership: |
|

SLC23A3
Genomic Map