Assay Details
| Gene Symbol: GPR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092160.2 | XP_014947646.1 | 3 - 4 | 2372 | 68 | |
| XM_027972737.1 | XP_027828538.1 | 2 - 3 | 2508 | 68 | ||
More Information
Set Membership: |
|

GPR61
Genomic Map