Assay Details
| Gene Symbol: NEMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093322.2 | XP_014948808.2 | 4 - 5 | 606 | 75 | |
| XM_027964705.1 | XP_027820506.1 | 4 - 5 | 3337 | 75 | ||
| XM_027964706.1 | XP_027820507.1 | 5 - 6 | 620 | 75 | ||
More Information
Set Membership: |
|

NEMP2
Genomic Map