Assay Details
Gene Symbol: LOC101121054 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003092.4 | XP_004003141.1 | 13 - 14 | 1948 | 86 | |
XM_027957697.1 | XP_027813498.1 | 12 - 13 | 1544 | 86 | ||
XM_027957708.1 | XP_027813509.1 | 11 - 12 | 1457 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
