Assay Details
| Gene Symbol: GLRX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013584.4 | XP_004013633.1 | 3 - 4 | 425 | 82 | |
| XM_012187004.3 | XP_012042394.1 | 3 - 4 | 426 | 82 | ||
| XM_012187005.3 | XP_012042395.1 | 3 - 4 | 425 | 82 | ||
| XM_027976264.1 | XP_027832065.1 | 3 - 4 | 306 | 82 | ||
More Information
Set Membership: |
|

GLRX2
Genomic Map