Assay Details
Gene Symbol: AURKAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013783.3 | XP_004013832.1 | 3 - 4 | 791 | 65 | |
XM_004013784.4 | XP_004013833.1 | 3 - 4 | 702 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
