Assay Details
Gene Symbol: FILIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097290.2 | XP_014952776.2 | 5 - 6 | 3900 | 79 | |
XM_027972285.1 | XP_027828086.1 | 5 - 6 | 3900 | 79 | ||
XM_027972286.1 | XP_027828087.1 | 6 - 7 | 3612 | 79 | ||
XM_027972287.1 | XP_027828088.1 | 5 - 6 | 3900 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
