Assay Details
Gene Symbol: TTC9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978330.1 | XP_027834131.1 | 1 - 2 | 2914 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
