Assay Details
| Gene Symbol: SLC9A3R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013137.4 | XP_004013186.2 | 4 - 5 | 1053 | 62 | |
| GenBank mRNA | DQ272258.1 | - | 2 - 3 | 197 | 62 | |
More Information
Set Membership: |
|

SLC9A3R1
Genomic Map