Assay Details
| Gene Symbol: RPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014418.3 | XP_004014467.1 | 5 - 6 | 340 | 62 | |
| XM_012188865.3 | XP_012044255.1 | 5 - 6 | 336 | 62 | ||
More Information
Set Membership: |
|

RPS21
Genomic Map