Assay Details
Gene Symbol: HSPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100102.2 | XP_014955588.2 | 7 - 8 | 1276 | 76 | |
XM_027978793.1 | XP_027834594.1 | 7 - 8 | 1273 | 76 | ||
XM_027978794.1 | XP_027834595.1 | 7 - 8 | 1277 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
