Assay Details
| Gene Symbol: NIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002683.3 | XP_004002732.1 | 3 - 4 | 435 | 67 | |
| XM_012184743.3 | XP_012040133.1 | 3 - 4 | 493 | 67 | ||
More Information
Set Membership: |
|

NIT1
Genomic Map