Assay Details
Gene Symbol: MED10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980179.1 | XP_027835980.1 | 2 - 3 | 272 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
